Search
Search

RAX2| Gene Symbol: RAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319074.1 | NP_001306003.1 | 3 | 1457 | ||
| NM_032753.3 | NP_116142.1 | 3 | 1198 | |||
| GenBank mRNA | AY211277.1 | - | 3 | 1190 | ||
| BC007284.1 | - | 4 | 1095 | 3344166 | ||
| BC018709.1 | - | 3 | 1457 | 4635722 | ||
| BC032512.1 | - | 3 | 1468 | 5491469 | ||