Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017564.9 | NP_060034.9 | 59 | 6608 | ||
XM_011538537.2 | XP_011536839.1 | 59 | 6649 | |||
XM_011538538.2 | XP_011536840.1 | 59 | 6649 | |||
XM_011538539.2 | XP_011536841.1 | 45 | 4852 | |||
XM_011538541.2 | XP_011536843.1 | 37 | 3959 | |||
XM_011538542.2 | XP_011536844.1 | 26 | 2768 | |||
XM_017019585.1 | XP_016875074.1 | 59 | 6649 | |||
GenBank mRNA | AB052958.1 | - | 59 | 6604 | ||
AF160476.1 | - | 14 | 1630 | |||
AJ295695.1 | - | 59 | 6608 | |||
AK024503.1 | - | 27 | 2950 | |||
AK074051.1 | - | 37 | 3979 | |||
AK160380.1 | - | 58 | 6326 | |||
AL133021.1 | - | 19 | 2090 | |||
AY227444.1 | - | 28 | 3017 | |||
AY311388.1 | - | 59 | 6626 |