Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197107.1 | NP_001184036.1 | 8 | 2081 | ||
NM_001197108.1 | NP_001184037.1 | 7 | 1424 | |||
NM_001197109.1 | NP_001184038.1 | 7 | 1141 | |||
NM_014575.3 | NP_055390.1 | 8 | 2120 | |||
GenBank mRNA | AF070614.1 | - | 7 | 1230 | ||
AF145713.1 | - | 8 | 1615 | |||
AF145714.1 | - | 7 | 919 | |||
AF145715.1 | - | 8 | 1576 | |||
AK091871.1 | - | 7 | 1062 | |||
BC005947.1 | - | 7 | 957 | 4246326 | ||
BC036535.2 | - | 8 | 2120 | 5263291 |
Gene Symbol: IQCJ-SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197113.1 | NP_001184042.1 | 11 | 1955 | ||
NM_001197114.1 | NP_001184043.1 | 10 | 1874 | |||
GenBank mRNA | AK096479.1 | - | 11 | 1861 |