Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPIP5K1 SIRNA Assays ›
Gene Symbol:
PPIP5K1
Gene Name:
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases:
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location:
Chr.15: 43533462 - 43590272 on Build GRCh38
UniGene ID:
Hs.156814
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21751
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPIP5K1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130858.2
NP_001124330.1
15
1679
NM_001130859.2
NP_001124331.1
14
1674
NM_001190214.1
NP_001177143.1
15
1679
NM_014659.5
NP_055474.3
14
1597
XM_005254803.1
XP_005254860.1
15
1679
XM_005254804.1
XP_005254861.1
15
1679
XM_006720786.2
XP_006720849.1
15
1679
XM_011522249.1
XP_011520551.1
15
1679
XM_011522250.1
XP_011520552.1
15
1782
XM_011522252.1
XP_011520554.1
14
1653
XM_011522254.1
XP_011520556.1
14
1549
XM_011522258.1
XP_011520560.1
9
939
XM_011522259.1
XP_011520561.1
15
1679
XM_017022735.1
XP_016878224.1
14
1996
XM_017022736.1
XP_016878225.1
14
1964
XM_017022737.1
XP_016878226.1
14
1645
XM_017022738.1
XP_016878227.1
14
1593
XM_017022739.1
XP_016878228.1
14
1593
XM_017022740.1
XP_016878229.1
15
1782
XM_017022741.1
XP_016878230.1
14
1995
XM_017022742.1
XP_016878231.1
15
1787
XM_017022743.1
XP_016878232.1
14
1653
XM_017022744.1
XP_016878233.1
14
1594
XM_017022745.1
XP_016878234.1
15
1782
XM_017022746.1
XP_016878235.1
14
1653
XM_017022747.1
XP_016878236.1
14
1593
XM_017022748.1
XP_016878237.1
14
1995
XM_017022749.1
XP_016878238.1
15
1755
XM_017022750.1
XP_016878239.1
14
1596
XM_017022751.1
XP_016878240.1
14
1549
XM_017022752.1
XP_016878241.1
15
1679
XM_017022753.1
XP_016878242.1
14
1653
XM_017022754.1
XP_016878243.1
15
1787
XM_017022755.1
XP_016878244.1
14
1653
XM_017022756.1
XP_016878245.1
14
1591
XM_017022757.1
XP_016878246.1
14
1549
XM_017022758.1
XP_016878247.1
9
939
XM_017022759.1
XP_016878248.1
14
1593
XM_017022760.1
XP_016878249.1
14
1653
GenBank mRNA
AB002375.1
-
14
1601
AF502586.1
-
14
1569
AF502587.1
-
14
1601
AF502588.1
-
14
1569
AF502589.1
-
14
1601
AF543190.1
-
14
1551
AK307438.1
-
15
1643
BC050263.1
-
13
1625
5521199
BC057395.1
-
14
1625
6142663
BX647814.1
-
15
1634
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
inositol metabolic process
phosphorylation
dephosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0