Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTSS1 SIRNA Assays ›
Gene Symbol:
MTSS1
Gene Name:
metastasis suppressor 1
Gene Aliases:
MIM, MIMA, MIMB
Chromosome Location:
Chr.8: 124550770 - 124728507 on Build GRCh38
UniGene ID:
Hs.336994
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21822
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTSS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282971.1
NP_001269900.1
15
2624
NM_001282974.1
NP_001269903.1
14
2537
NM_014751.5
NP_055566.3
14
2612
XM_005251111.1
XP_005251168.1
14
2917
XM_005251113.1
XP_005251170.1
13
2722
XM_005251118.1
XP_005251175.1
14
2395
XM_006716700.1
XP_006716763.1
14
2914
XM_006716701.1
XP_006716764.1
13
2905
XM_006716702.1
XP_006716765.1
14
2866
XM_006716703.1
XP_006716766.1
13
2671
XM_006716704.1
XP_006716767.1
15
2587
XM_006716705.1
XP_006716768.1
15
2539
XM_006716706.1
XP_006716769.1
14
2344
XM_011517403.1
XP_011515705.1
13
2371
XM_011517404.2
XP_011515706.1
12
14083
XM_011517407.1
XP_011515709.1
5
1571
XM_017014086.1
XP_016869575.1
13
2902
XM_017014087.1
XP_016869576.1
14
2842
XM_017014088.1
XP_016869577.1
13
2830
XM_017014089.1
XP_016869578.1
13
2827
XM_017014090.1
XP_016869579.1
12
3463
XM_017014091.1
XP_016869580.1
14
2351
XM_017014092.1
XP_016869581.1
12
2710
XM_017014093.1
XP_016869582.1
14
2575
XM_017014094.1
XP_016869583.1
14
2527
XM_017014095.1
XP_016869584.1
15
2515
XM_017014096.1
XP_016869585.1
15
2512
XM_017014097.1
XP_016869586.1
14
2500
XM_017014098.1
XP_016869587.1
13
2383
XM_017014099.1
XP_016869588.1
13
2380
XM_017014100.1
XP_016869589.1
13
2332
XM_017014101.1
XP_016869590.1
12
2162
GenBank mRNA
AB007889.2
-
11
1794
AF086645.1
-
14
2523
AK027015.1
-
15
2390
AK126651.1
-
2
1324
AK126989.1
-
7
1368
AK295681.1
-
7
1423
AK302036.1
-
8
1868
AK310792.1
-
6
1334
BC023998.1
-
8
1384
4826982
EF560715.1
-
14
2168
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
movement of cell or subcellular component
plasma membrane organization
cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
microspike assembly
actin cytoskeleton organization
negative regulation of epithelial cell proliferation
renal tubule morphogenesis
cellular response to fluid shear stress
glomerulus morphogenesis
nephron tubule epithelial cell differentiation
xenophagy
epithelial cell proliferation involved in renal tubule morphogenesis
actin monomer binding
receptor binding
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0