Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008527.1 | NP_001008527.1 | 9 | 838 | ||
XM_006245004.2 | XP_006245066.1 | 9 | 866 | |||
XM_008767121.1 | XP_008765343.1 | 9 | 867 | |||
XM_008767122.1 | XP_008765344.1 | 5 | 1535 | |||
XM_008767123.1 | XP_008765345.1 | 3 | 357 | |||
XM_008767124.1 | XP_008765346.1 | 4 | 533 | |||
GenBank mRNA | AY665586.1 | - | 9 | 838 |