Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTSS1 SIRNA Assays ›
Gene Symbol:
MTSS1
Gene Name:
metastasis suppressor 1
Gene Aliases:
MIM, MIMA, MIMB
Chromosome Location:
Chr.8: 124550770 - 124728507 on Build GRCh38
UniGene ID:
Hs.336994
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21918
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTSS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282971.1
NP_001269900.1
15
2636
NM_001282974.1
NP_001269903.1
14
2549
NM_014751.5
NP_055566.3
14
2624
XM_005251111.1
XP_005251168.1
14
2929
XM_005251113.1
XP_005251170.1
13
2734
XM_005251118.1
XP_005251175.1
14
2407
XM_006716700.1
XP_006716763.1
14
2926
XM_006716701.1
XP_006716764.1
13
2917
XM_006716702.1
XP_006716765.1
14
2878
XM_006716703.1
XP_006716766.1
13
2683
XM_006716704.1
XP_006716767.1
15
2599
XM_006716705.1
XP_006716768.1
15
2551
XM_006716706.1
XP_006716769.1
14
2356
XM_011517403.1
XP_011515705.1
13
2383
XM_011517404.2
XP_011515706.1
12
14095
XM_011517407.1
XP_011515709.1
5
1583
XM_017014086.1
XP_016869575.1
13
2914
XM_017014087.1
XP_016869576.1
14
2854
XM_017014088.1
XP_016869577.1
13
2842
XM_017014089.1
XP_016869578.1
13
2839
XM_017014090.1
XP_016869579.1
12
3475
XM_017014091.1
XP_016869580.1
14
2363
XM_017014092.1
XP_016869581.1
12
2722
XM_017014093.1
XP_016869582.1
14
2587
XM_017014094.1
XP_016869583.1
14
2539
XM_017014095.1
XP_016869584.1
15
2527
XM_017014096.1
XP_016869585.1
15
2524
XM_017014097.1
XP_016869586.1
14
2512
XM_017014098.1
XP_016869587.1
13
2395
XM_017014099.1
XP_016869588.1
13
2392
XM_017014100.1
XP_016869589.1
13
2344
XM_017014101.1
XP_016869590.1
12
2174
GenBank mRNA
AB007889.2
-
11
1806
AF086645.1
-
14
2535
AK027015.1
-
15
2402
AK126651.1
-
2
1336
AK126989.1
-
7
1380
AK295681.1
-
7
1435
AK302036.1
-
8
1880
AK310792.1
-
6
1346
BC023998.1
-
8
1396
4826982
EF560715.1
-
14
2180
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
movement of cell or subcellular component
plasma membrane organization
cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
microspike assembly
actin cytoskeleton organization
negative regulation of epithelial cell proliferation
renal tubule morphogenesis
cellular response to fluid shear stress
glomerulus morphogenesis
nephron tubule epithelial cell differentiation
xenophagy
epithelial cell proliferation involved in renal tubule morphogenesis
actin monomer binding
receptor binding
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0