Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 32 | 5827 | ||
NM_001265611.1 | NP_001252540.1 | 30 | 4978 | |||
NM_014994.2 | NP_055809.2 | 31 | 5809 | |||
NR_049761.1 | 30 | 5619 | ||||
NR_049762.1 | 29 | 5570 | ||||
XM_006720438.1 | XP_006720501.1 | 30 | 5626 | |||
XM_006720439.2 | XP_006720502.1 | 18 | 3901 | |||
XM_011521383.1 | XP_011519685.1 | 31 | 5644 | |||
XM_017022017.1 | XP_016877506.1 | 30 | 5920 | |||
GenBank mRNA | AB011168.2 | - | 32 | 5737 | ||
AB177850.1 | - | 23 | 4651 | |||
AL833267.1 | - | 31 | 5729 | |||
BC113983.1 | - | 30 | 4870 | 40083186 | ||
BC114493.1 | - | 32 | 5719 | 40083182 | ||
BX648786.1 | - | 31 | 5760 |