Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCP110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199022.2 | NP_001185951.2 | 10, 11 | 2951 | ||
NM_001323569.1 | NP_001310498.1 | 11, 12 | 3094 | |||
NM_001323570.1 | NP_001310499.1 | 12, 13 | 3539 | |||
NM_001323571.1 | NP_001310500.1 | 12, 13 | 3334 | |||
NM_001323572.1 | NP_001310501.1 | 10, 11 | 2951 | |||
NM_001323576.1 | NP_001310505.1 | 11, 12 | 3094 | |||
NM_001323577.1 | NP_001310506.1 | 10, 11 | 2951 | |||
NM_014711.5 | NP_055526.4 | 11, 12 | 3094 | |||
XM_011545991.2 | XP_011544293.1 | 12, 13 | 3517 | |||
XM_011545992.2 | XP_011544294.1 | 12, 13 | 3235 | |||
XM_017023908.1 | XP_016879397.1 | 11, 12 | 4011 | |||
GenBank mRNA | AB007879.1 | - | 11, 12 | 2995 | ||
AK300426.1 | - | 10, 11 | 2864 | |||
BC036654.2 | - | 11, 12 | 3065 | 5267904 | ||
CN307903.1 | - | 3, 4 | 238 | |||
CR749255.1 | - | 10, 11 | 2851 | |||
DA370828.1 | - | 5, 6 | 517 |