Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003820.1 | NP_001003820.1 | 1 | 233 | ||
NM_001134912.1 | NP_001128384.1 | 1 | 267 | |||
GenBank mRNA | AY567970.1 | - | 1 | 233 | ||
EU152989.1 | - | 1 | 267 |