Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271779.1 | NP_001258708.1 | 3 | 579 | ||
NM_001271780.1 | NP_001258709.1 | 2 | 294 | |||
NM_001271781.1 | NP_001258710.1 | 2 | 169 | |||
NM_014814.2 | NP_055629.1 | 2 | 420 | |||
XM_005265619.1 | XP_005265676.1 | 2 | 516 | |||
XM_011534288.1 | XP_011532590.1 | 3 | 475 | |||
XM_017007569.1 | XP_016863058.1 | 3 | 322 | |||
GenBank mRNA | AF215935.1 | - | 2 | 286 | ||
AF530062.1 | - | 2 | 233 | |||
AK095059.1 | - | 1 | 1002 | |||
AK290205.1 | - | 2 | 274 | |||
AL529900.3 | - | 2 | 155 | |||
AY359879.1 | - | 2 | 266 | |||
AY568086.1 | - | 2 | 258 | |||
BC000630.1 | - | 2 | 250 | 3344973 | ||
BC000904.2 | - | 2 | 271 | 2900971 | ||
BC012369.1 | - | 2 | 268 | 3858094 | ||
BG723464.1 | - | 2 | 297 | |||
BX647742.1 | - | 3 | 446 | |||
CD692691.1 | - | 3 | 479 | |||
D14663.1 | - | 2 | 258 | |||
HY010377.1 | - | 2 | 421 |