Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Knstrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004264.1 | NP_001004264.1 | 9 | 998 | ||
XM_006234757.2 | XP_006234819.1 | 10 | 1089 | |||
XM_006234758.2 | XP_006234820.1 | 9 | 855 | |||
GenBank mRNA | BC079438.1 | - | 9 | 998 | 7115344 |