Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZC3H11A SIRNA Assays ›
          Gene Symbol:
          ZC3H11A LOC441155
          Gene Name:
          zinc finger CCCH-type containing 11A
          zinc finger CCCH-type domain-containing-like
          Gene Aliases:
          ZC3HDC11A
          -
          Chromosome Location:
          Chr.1: 203795537 - 203854128 on Build GRCh38
          Chromosome Location:
          Chr.6: 65301418 - 65306069 on Build GRCh38
          UniGene ID:
          Hs.532399
          Hs.546686
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 22067

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZC3H11A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001319238.1 NP_001306167.1 6 1291
          NM_001319239.1 NP_001306168.1 6 1186
          NM_014827.5 NP_055642.3 6 862
          XM_005245643.2 XP_005245700.1 6 881
          XM_005245644.3 XP_005245701.1 5 565
          XM_006711673.2 XP_006711736.1 6 728
          XM_011510199.1 XP_011508501.1 4 1933
          XM_011510200.1 XP_011508502.1 5 953
          XM_011510201.1 XP_011508503.1 6 1139
          XM_011510202.1 XP_011508504.1 4 878
          XM_017002956.1 XP_016858445.1 5 2187
          XM_017002957.1 XP_016858446.1 5 2039
          XM_017002958.1 XP_016858447.1 5 2026
          XM_017002959.1 XP_016858448.1 6 1099
          XM_017002960.1 XP_016858449.1 5 2187
          XM_017002961.1 XP_016858450.1 4 1933
          XM_017002962.1 XP_016858451.1 5 953
          XM_017002963.1 XP_016858452.1 6 745
          XM_017002964.1 XP_016858453.1 4 1933
          XM_017002965.1 XP_016858454.1 3 1262
          GenBank mRNA AB014563.1 - 3 373
          AK095858.1 - 6 1160
          AK296983.1 - 2 220
          BC014268.2 - 6 700 3844154
          BC038513.2 - 6 1017
          BX648271.1 - 3 3675
          BX649148.1 - 3 4476
          CR627439.1 - 4 1883
          CR627446.1 - 3 3986
          Gene Symbol: LOC441155
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271675.1 NP_001258604.1 2 894
          XM_017010889.1 XP_016866378.1 2 410
          GenBank mRNA AK302514.1 - 2 850

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          termination of RNA polymerase II transcription
          RNA export from nucleus
          mRNA export from nucleus
          poly(A)+ mRNA export from nucleus
          mRNA 3'-end processing
          protein binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0