Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFASC SIRNA Assays ›
Gene Symbol:
NFASC
Gene Name:
neurofascin
Gene Aliases:
NF, NRCAML
Chromosome Location:
Chr.1: 204828652 - 205022822 on Build GRCh38
UniGene ID:
Hs.13349
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
220925
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFASC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005388.2
NP_001005388.2
3
406
NM_001005389.1
NP_001005389.2
3
406
NM_001160331.1
NP_001153803.1
1
168
NM_001160332.1
NP_001153804.1
3
406
NM_001160333.1
NP_001153805.1
3
406
NM_015090.3
NP_055905.2
3
406
XM_005244989.3
XP_005245046.2
3
486
XM_005244991.3
XP_005245048.2
3
486
XM_005244992.4
XP_005245049.2
3
486
XM_005244993.3
XP_005245050.2
3
486
XM_005244997.3
XP_005245054.2
3
486
XM_011509311.1
XP_011507613.1
1
168
XM_011509312.1
XP_011507614.1
3
486
XM_011509313.1
XP_011507615.1
1
168
XM_011509314.1
XP_011507616.1
3
486
XM_011509315.1
XP_011507617.1
1
168
XM_011509316.1
XP_011507618.1
3
365
XM_011509317.1
XP_011507619.1
1
168
XM_011509318.2
XP_011507620.1
3
408
XM_011509319.1
XP_011507621.1
3
365
XM_011509320.2
XP_011507622.1
3
408
XM_011509321.1
XP_011507623.1
1
168
XM_011509322.1
XP_011507624.1
1
168
XM_011509323.1
XP_011507625.1
1
168
XM_011509324.1
XP_011507626.1
1
168
XM_011509325.1
XP_011507627.1
1
168
XM_011509326.1
XP_011507628.1
1
168
XM_011509327.1
XP_011507629.1
1
168
XM_011509328.1
XP_011507630.1
1
168
XM_017000733.1
XP_016856222.1
3
486
XM_017000734.1
XP_016856223.1
3
486
XM_017000735.1
XP_016856224.1
1
168
XM_017000736.1
XP_016856225.1
1
168
XM_017000737.1
XP_016856226.1
1
168
XM_017000738.1
XP_016856227.1
3
408
XM_017000739.1
XP_016856228.1
3
408
XM_017000740.1
XP_016856229.1
3
408
XM_017000741.1
XP_016856230.1
1
40
XM_017000742.1
XP_016856231.1
1
168
XM_017000743.1
XP_016856232.1
3
408
GenBank mRNA
AB018299.3
-
2
237
AK090639.1
-
3
365
AK097802.1
-
216
AK295291.1
-
3
385
AK299262.1
-
3
385
AK308686.1
-
3
486
BC040674.1
-
3
411
BC117674.2
-
2
255
BC137013.1
-
1
143
9021006
BC144454.1
-
1
143
9052980
BC144455.1
-
1
143
BG702675.1
-
3
411
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein localization to paranode region of axon
axon guidance
peripheral nervous system development
transmission of nerve impulse
paranodal junction assembly
heterotypic cell-cell adhesion
myelination
clustering of voltage-gated sodium channels
synapse organization
protein localization to juxtaparanode region of axon
protein targeting to plasma membrane
protein binding
protein domain specific binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-68wbn:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0