Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289522.1 | NP_001276451.1 | 17 | 2600 | ||
NM_001289523.1 | NP_001276452.1 | 17 | 2600 | |||
NM_023525.2 | NP_076014.1 | 17 | 2600 | |||
GenBank mRNA | BC053097.1 | - | 17 | 2569 | ||
BC060119.1 | - | 17 | 2567 | |||
BC060717.1 | - | 8 | 1057 | |||
BC137856.1 | - | 17 | 2452 | 8860875 | ||
BC144972.1 | - | 17 | 2452 | 9053520 | ||
BC144973.1 | - | 17 | 2452 |