Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAC1 SIRNA Assays ›
          Gene Symbol:
          Rac1
          Gene Name:
          ras-related C3 botulinum toxin substrate 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 13090338 - 13111835 on Build Rnor_6.0
          UniGene ID:
          Rn.29157
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 221515

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rac1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_134366.1 NP_599193.1 6 724
          XM_008768986.1 XP_008767208.1 7 815
          GenBank mRNA AY491395.1 - 6 724

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          auditory receptor cell morphogenesis
          mast cell chemotaxis
          epithelial cell morphogenesis
          endocytosis
          phagocytosis, engulfment
          chemotaxis
          hyperosmotic response
          cytoskeleton organization
          actin filament organization
          cell adhesion
          G-protein coupled receptor signaling pathway
          small GTPase mediated signal transduction
          axon guidance
          metabolic process
          cell proliferation
          positive regulation of lamellipodium assembly
          positive regulation of cell-substrate adhesion
          regulation of neuron maturation
          dendrite development
          cell migration
          Rac protein signal transduction
          cerebral cortex radially oriented cell migration
          embryonic olfactory bulb interneuron precursor migration
          cerebral cortex GABAergic interneuron development
          regulation of cell morphogenesis
          lamellipodium assembly
          actin cytoskeleton organization
          actin filament polymerization
          regulation of cell migration
          positive regulation of actin filament polymerization
          ruffle organization
          negative regulation of interleukin-23 production
          substrate adhesion-dependent cell spreading
          positive regulation of phosphatidylinositol 3-kinase activity
          engulfment of apoptotic cell
          cell-cell junction organization
          bone resorption
          positive regulation of DNA replication
          anatomical structure arrangement
          dendrite morphogenesis
          cell motility
          homeostasis of number of cells within a tissue
          positive regulation of stress fiber assembly
          localization within membrane
          positive regulation of focal adhesion assembly
          synaptic transmission, GABAergic
          Wnt signaling pathway, planar cell polarity pathway
          regulation of respiratory burst
          cellular response to mechanical stimulus
          semaphorin-plexin signaling pathway
          dopaminergic neuron differentiation
          protein localization to plasma membrane
          positive regulation of neutrophil chemotaxis
          cochlea morphogenesis
          ruffle assembly
          positive regulation of substrate adhesion-dependent cell spreading
          GTPase activity
          protein binding
          GTP binding
          Rab GTPase binding
          enzyme binding
          protein kinase binding
          GTP-dependent protein binding
          thioesterase binding
          histone deacetylase binding
          Rho GDP-dissociation inhibitor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0