Search Thermo Fisher Scientific
Gene Symbol: H2BFWT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002916.4 | NP_001002916.3 | 3 | 660 | ||
GenBank mRNA | BC038109.2 | - | 3 | 657 | 5266336 | |
BC118604.1 | - | 3 | 653 | 40109007 | ||
BC121816.1 | - | 3 | 653 | 40109006 |