Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAPVD1 SIRNA Assays ›
Gene Symbol:
GAPVD1
Gene Name:
GTPase activating protein and VPS9 domains 1
Gene Aliases:
GAPEX5, GAPex-5, RAP6
Chromosome Location:
Chr.9: 125261794 - 125365011 on Build GRCh38
UniGene ID:
Hs.495134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
222689
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAPVD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282679.1
NP_001269608.1
28
5722
NM_001282680.1
NP_001269609.1
28
5668
NM_001282681.1
NP_001269610.1
25
5273
NM_015635.3
NP_056450.2
27
5583
XM_005251901.3
XP_005251958.1
27
5532
XM_005251904.3
XP_005251961.1
26
5448
XM_006717044.3
XP_006717107.1
27
5454
XM_011518499.2
XP_011516801.1
28
5627
XM_011518500.2
XP_011516802.1
29
5744
XM_011518502.2
XP_011516804.1
28
5555
XM_011518506.2
XP_011516808.1
28
5552
XM_011518507.2
XP_011516809.1
27
5551
XM_017014600.1
XP_016870089.1
27
5437
XM_017014601.1
XP_016870090.1
26
5459
XM_017014602.1
XP_016870091.1
28
5679
XM_017014603.1
XP_016870092.1
28
5626
XM_017014604.1
XP_016870093.1
29
5728
XM_017014605.1
XP_016870094.1
26
5459
XM_017014606.1
XP_016870095.1
28
5573
XM_017014607.1
XP_016870096.1
28
5582
XM_017014608.1
XP_016870097.1
26
5392
XM_017014609.1
XP_016870098.1
27
5560
XM_017014610.1
XP_016870099.1
25
5378
XM_017014611.1
XP_016870100.1
26
5371
GenBank mRNA
AK023841.1
-
19
3616
AL834325.1
-
7
1913
BC114937.1
-
28
5551
40083103
BC114962.1
-
25
5306
40083104
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
signal transduction
positive regulation of GTPase activity
regulation of protein transport
cell-cell adhesion
guanyl-nucleotide exchange factor activity
GTPase activator activity
GTPase activating protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-jct7h:80/100.66.72.173:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0