Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RALGAPA1 SIRNA Assays ›
          Gene Symbol:
          RALGAPA1 RALGAPA1P1
          Gene Name:
          Ral GTPase activating protein catalytic alpha subunit 1
          Ral GTPase activating protein catalytic alpha subunit 1 pseudogene 1
          Gene Aliases:
          GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
          GARNL2, GARNL2P, RALGAPA1P, bA235C23.1
          Chromosome Location:
          Chr.14: 35538352 - 35809304 on Build GRCh38
          Chromosome Location:
          Chr.9: 105519824 - 105527319 on Build GRCh38
          UniGene ID:
          Hs.113150
          Hs.113150
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 222711

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RALGAPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001283043.1 NP_001269972.1 2, 3 616
          NM_001283044.1 NP_001269973.1 2, 3 616
          NM_014990.1 NP_055805.1 2, 3 616
          NM_194301.2 NP_919277.2 2, 3 616
          XM_005267491.3 XP_005267548.1 2, 3 694
          XM_005267492.3 XP_005267549.1 2, 3 694
          XM_005267493.3 XP_005267550.1 2, 3 694
          XM_006720098.2 XP_006720161.1 2, 3 694
          XM_006720099.2 XP_006720162.1 2, 3 694
          XM_006720100.3 XP_006720163.1 2, 3 694
          XM_006720101.2 XP_006720164.1 2, 3 694
          XM_006720102.3 XP_006720165.1 2, 3 400
          XM_006720103.2 XP_006720166.1 2, 3 694
          XM_006720104.3 XP_006720167.1 2, 3 694
          XM_011536615.2 XP_011534917.1 2, 3 694
          XM_011536616.2 XP_011534918.1 2, 3 694
          XM_017021141.1 XP_016876630.1 2, 3 694
          XM_017021142.1 XP_016876631.1 2, 3 694
          XM_017021143.1 XP_016876632.1 2, 3 694
          XM_017021144.1 XP_016876633.1 2, 3 694
          XM_017021145.1 XP_016876634.1 2, 3 694
          GenBank mRNA AB511280.1 - 2, 3 225
          AY596970.1 - 2, 3 616
          AY596971.1 - 2, 3 616
          BC150596.1 - 2, 3 324 9056966
          Gene Symbol: RALGAPA1P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_104269.1 1 656

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: BE100000.
          This siRNA also targets the following transcripts in Mouse: NM_001003719, NM_001112714, NM_001286263, NM_019994, BC145119, and others.

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          regulation of small GTPase mediated signal transduction
          activation of GTPase activity
          GTPase activator activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0