Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213733.2 | NP_998898.1 | 7, 8 | 977 | ||
GenBank mRNA | AK150313.1 | - | 7, 8 | 978 | ||
AK156136.1 | - | 7, 8 | 978 | |||
AK170509.1 | - | 7, 8 | 978 | |||
BC023239.1 | - | 6, 7 | 655 | |||
BC055448.1 | - | 6, 7 | 652 | |||
BC069933.1 | - | 7, 8 | 972 | 4008091 | ||
BC080707.1 | - | 7, 8 | 850 |