Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 16 | 4767 | ||
XM_005261405.2 | XP_005261462.1 | 16 | 4767 | |||
XM_006724171.3 | XP_006724234.1 | 15 | 13545 | |||
XM_011530018.2 | XP_011528320.1 | 16 | 4946 | |||
XM_017028673.1 | XP_016884162.1 | 16 | 4740 | |||
GenBank mRNA | AB028966.3 | - | 11 | 3560 | ||
AB665284.1 | - | 16 | 4715 | |||
BC016465.1 | - | 8 | 1311 | 4154050 |