Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMAD3 SIRNA Assays ›
          Gene Symbol:
          SMAD3
          Gene Name:
          SMAD family member 3
          Gene Aliases:
          HSPC193, HsT17436, JV15-2, LDS1C, LDS3, MADH3
          Chromosome Location:
          Chr.15: 67065698 - 67195195 on Build GRCh38
          UniGene ID:
          Hs.727986
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 22903

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMAD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145102.1 NP_001138574.1 9 5220
          NM_001145103.1 NP_001138575.1 9 5031
          NM_001145104.1 NP_001138576.1 7 4664
          NM_005902.3 NP_005893.1 9 5455
          XM_011521559.2 XP_011519861.1 8 5485
          XM_011521560.1 XP_011519862.1 9 5397
          GenBank mRNA AF151027.1 - 1 704
          AL110265.1 - 1 1258
          BC000414.1 - 1 608
          BC007496.1 - 1 608
          CR749287.1 - 1 4215

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          response to hypoxia
          in utero embryonic development
          mesoderm formation
          somitogenesis
          liver development
          heart looping
          osteoblast development
          immune system development
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          immune response
          cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          SMAD protein complex assembly
          endoderm development
          embryonic pattern specification
          positive regulation of gene expression
          positive regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          regulation of striated muscle tissue development
          regulation of transforming growth factor beta receptor signaling pathway
          evasion or tolerance of host defenses by virus
          signal transduction involved in regulation of gene expression
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of bone mineralization
          negative regulation of transforming growth factor beta receptor signaling pathway
          thyroid gland development
          primary miRNA processing
          positive regulation of chondrocyte differentiation
          positive regulation of interleukin-1 beta production
          regulation of transforming growth factor beta2 production
          positive regulation of transforming growth factor beta3 production
          activin receptor signaling pathway
          negative regulation of osteoblast proliferation
          nodal signaling pathway
          wound healing
          T cell activation
          positive regulation of transcription factor import into nucleus
          negative regulation of apoptotic process
          cell-cell junction organization
          negative regulation of fat cell differentiation
          negative regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          paraxial mesoderm morphogenesis
          developmental growth
          embryonic foregut morphogenesis
          embryonic cranial skeleton morphogenesis
          regulation of epithelial cell proliferation
          negative regulation of inflammatory response
          regulation of immune response
          positive regulation of positive chemotaxis
          regulation of binding
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          pericardium development
          transdifferentiation
          SMAD protein signal transduction
          negative regulation of wound healing
          lens fiber cell differentiation
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of extracellular matrix assembly
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          core promoter proximal region sequence-specific DNA binding
          transcription factor activity, protein binding
          RNA polymerase II activating transcription factor binding
          transcription factor activity, sequence-specific DNA binding
          transforming growth factor beta receptor binding
          protein binding
          collagen binding
          transcription factor binding
          zinc ion binding
          protein kinase binding
          phosphatase binding
          transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
          chromatin DNA binding
          ubiquitin protein ligase binding
          enhancer binding
          identical protein binding
          protein homodimerization activity
          ubiquitin binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          co-SMAD binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rln4r:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0