Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PAMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001991.2 | NP_001001991.1 | 2 | 663 | ||
NM_001282675.1 | NP_001269604.1 | 4 | 653 | |||
NM_001282676.1 | NP_001269605.1 | 2 | 663 | |||
NM_015430.3 | NP_056245.2 | 2 | 663 | |||
GenBank mRNA | AF370388.1 | - | 1 | 151 | ||
AK027841.1 | - | 2 | 263 | |||
AK297092.1 | - | 4 | 654 | |||
AK300185.1 | - | 2 | 248 | |||
AK316188.1 | - | 2 | 235 | |||
AL832391.1 | - | 2 | 229 | |||
AY358346.1 | - | 2 | 242 | |||
BC089434.1 | - | 2 | 255 | 30558617 | ||
BX640676.1 | - | 2 | 663 |