Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WDPCP SIRNA Assays ›
          Gene Symbol:
          WDPCP
          Gene Name:
          WD repeat containing planar cell polarity effector
          Gene Aliases:
          BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
          Chromosome Location:
          Chr.2: 63120755 - 63840825 on Build GRCh38
          UniGene ID:
          Hs.414952
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 23286

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WDPCP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042692.2 NP_001036157.1 3 432
          NM_015910.5 NP_056994.3 9 1247
          NR_122106.1 3 432
          XM_005264348.3 XP_005264405.1 9 1287
          XM_011532881.2 XP_011531183.1 10 1071
          XM_011532884.2 XP_011531186.1 9 1287
          XM_011532886.2 XP_011531188.1 9 1287
          XM_011532887.2 XP_011531189.1 9 1287
          XM_011532890.2 XP_011531192.1 9 1287
          XM_011532891.2 XP_011531193.1 10 1272
          XM_017004251.1 XP_016859740.1 11 1925
          XM_017004252.1 XP_016859741.1 10 1071
          XM_017004253.1 XP_016859742.1 9 1287
          XM_017004254.1 XP_016859743.1 9 1287
          GenBank mRNA AF131737.1 - 3 432
          AK001849.1 - 1 147
          AK022647.1 - 3 406
          AK055030.1 - 10 1272
          AK309028.1 - 4 487
          AK310290.1 - 9 914
          AL833623.1 - 9 1033
          BC093752.1 - 3 377 7939597
          BC093754.1 - 3 377 7939599
          BX538331.1 - 9 993
          BX647132.1 - 3 3760

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          establishment of planar polarity
          kidney development
          auditory receptor cell morphogenesis
          smoothened signaling pathway
          regulation of fibroblast migration
          regulation of embryonic cell shape
          septin cytoskeleton organization
          regulation of protein localization
          cilium assembly
          embryonic digit morphogenesis
          camera-type eye development
          establishment of protein localization
          regulation of focal adhesion assembly
          digestive system development
          palate development
          cilium morphogenesis
          respiratory system development
          cardiovascular system development
          glomerular visceral epithelial cell migration
          regulation of ruffle assembly
          regulation of establishment of cell polarity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-tgh8t:80/100.66.77.52:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0