Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243473.2 | NP_001230402.1 | 6 | 1038 | ||
NM_001321214.1 | NP_001308143.1 | 7 | 892 | |||
NM_001321215.1 | NP_001308144.1 | 6 | 1131 | |||
NM_015681.4 | NP_056496.1 | 7 | 803 | |||
XM_005256607.2 | XP_005256664.1 | 7 | 809 | |||
GenBank mRNA | AB030506.1 | - | 7 | 792 | ||
AK293815.1 | - | 6 | 1104 |