Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000368.4 | NP_000359.1 | 23 | 5199 | ||
NM_001162426.1 | NP_001155898.1 | 23 | 5196 | |||
NM_001162427.1 | NP_001155899.1 | 22 | 5046 | |||
XM_005272211.1 | XP_005272268.1 | 23 | 5244 | |||
XM_006717271.1 | XP_006717334.1 | 23 | 5179 | |||
XM_011518979.2 | XP_011517281.1 | 23 | 5193 | |||
XM_017015096.1 | XP_016870585.1 | 24 | 5658 | |||
XM_017015097.1 | XP_016870586.1 | 24 | 5629 | |||
XM_017015098.1 | XP_016870587.1 | 23 | 5241 | |||
XM_017015099.1 | XP_016870588.1 | 22 | 5337 | |||
XM_017015100.1 | XP_016870589.1 | 22 | 5308 | |||
XM_017015101.1 | XP_016870590.1 | 22 | 5335 | |||
GenBank mRNA | AF013168.1 | - | 23 | 5185 | ||
D87683.1 | - | 10 | 3570 |