Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CERCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286760.1 | NP_001273689.1 | 3 | 283 | ||
NM_016174.4 | NP_057258.3 | 3 | 759 | |||
XM_005252035.1 | XP_005252092.1 | 5 | 1669 | |||
XM_011518762.1 | XP_011517064.1 | 6 | 1826 | |||
XM_011518763.2 | XP_011517065.1 | 3 | 1377 | |||
XM_017014794.1 | XP_016870283.1 | 7 | 1568 | |||
GenBank mRNA | AB040935.1 | - | 4 | 3272 | ||
AF177203.1 | - | 2 | 127 | |||
AK074519.1 | - | 2 | 195 | |||
AK074523.1 | - | 2 | 169 | |||
AK297929.1 | - | 2 | 255 | |||
AK314294.1 | - | 3 | 330 | |||
BC098432.1 | - | 3 | 357 | 6672104 | ||
BC108698.1 | - | 3 | 370 | |||
BC119698.1 | - | 3 | 232 | 40116832 | ||
BC119699.1 | - | 3 | 232 | 40116834 | ||
BX358794.2 | - | 3 | 408 | |||
CX871555.1 | - | 3 | 759 |