Search
Search

GNB5| Gene Symbol: GNB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006578.3 | NP_006569.1 | 3 | 352 | ||
| NM_016194.3 | NP_057278.2 | 5 | 457 | |||
| XM_011521162.2 | XP_011519464.1 | 3 | 709 | |||
| XM_011521163.2 | XP_011519465.1 | 3 | 234 | |||
| GenBank mRNA | AF017656.1 | - | 3 | 365 | ||
| AF300650.1 | - | 4 | 392 | |||
| AF501885.1 | - | 3 | 266 | |||
| AK314775.1 | - | 3 | 310 | |||
| AL117471.1 | - | 3 | 274 | |||
| AW967075.1 | - | 5 | 457 | |||
| BC011671.2 | - | 3 | 287 | |||
| BC013997.2 | - | 3 | 349 | 3621195 | ||