Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 33 | 4916 | ||
NM_001289397.1 | NP_001276326.1 | 32 | 4701 | |||
NM_001289398.1 | NP_001276327.1 | 6 | 883 | |||
NM_001289399.1 | NP_001276328.1 | 7 | 870 | |||
NM_001289400.1 | NP_001276329.1 | 7 | 1291 | |||
NM_003070.4 | NP_003061.3 | 33 | 4929 | |||
NM_139045.3 | NP_620614.2 | 32 | 4875 | |||
GenBank mRNA | AK094076.1 | - | 7 | 1291 | ||
AK298045.1 | - | 7 | 870 | |||
AK300093.1 | - | 8 | 935 | |||
AK308941.1 | - | 6 | 1028 | |||
CD107405.1 | - | 5 | 638 | |||
CD513892.1 | - | 5 | 679 | |||
X72889.1 | - | 33 | 4917 |