Search Thermo Fisher Scientific
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 15 | 1678 | ||
NM_001291721.1 | NP_001278650.1 | 14 | 1600 | |||
NM_001291722.1 | NP_001278651.1 | 15 | 1678 | |||
NM_014376.3 | NP_055191.2 | 15 | 1804 | |||
XM_011534516.2 | XP_011532818.1 | 15 | 1944 | |||
XM_017009341.1 | XP_016864830.1 | 15 | 1793 | |||
XM_017009342.1 | XP_016864831.1 | 16 | 1715 | |||
GenBank mRNA | AB032994.2 | - | 15 | 1618 | ||
AF160973.1 | - | 15 | 1587 | |||
AK223604.1 | - | 14 | 1556 | |||
AK294220.1 | - | 10 | 1047 | |||
AK295865.1 | - | 12 | 1227 | |||
AK303702.1 | - | 14 | 1553 | |||
AL136549.1 | - | 15 | 1677 | |||
AL161999.1 | - | 6 | 563 | |||
BC011762.1 | - | 15 | 1804 | 3619680 | ||
BC021008.1 | - | 15 | 1804 | |||
L47738.1 | - | 4 | 397 |