Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24103
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025068.1
NP_001020239.1
4
430
NM_001025069.1
NP_001020240.1
4
439
NM_001267616.1
NP_001254545.1
4
1586
NM_001267617.1
NP_001254546.1
4
396
NM_001267618.1
NP_001254547.1
3
527
NM_001267619.1
NP_001254548.1
3
361
NM_016300.4
NP_057384.2
4
413
NM_198399.1
NP_938409.1
3
421
XM_005264811.2
XP_005264868.1
4
442
XM_006712943.2
XP_006713006.1
4
442
XM_006712944.2
XP_006713007.1
4
442
XM_011533298.2
XP_011531600.1
4
442
XM_011533299.2
XP_011531601.1
4
986
XM_011533300.2
XP_011531602.1
4
605
XM_011533301.2
XP_011531603.1
4
436
XM_011533302.2
XP_011531604.1
5
528
XM_011533303.2
XP_011531605.1
5
1078
XM_017005574.1
XP_016861063.1
4
532
XM_017005575.1
XP_016861064.1
5
12165
XM_017005576.1
XP_016861065.1
4
986
XM_017005577.1
XP_016861066.1
4
436
XM_017005578.1
XP_016861067.1
4
412
XM_017005579.1
XP_016861068.1
4
986
XM_017005580.1
XP_016861069.1
5
1078
XM_017005581.1
XP_016861070.1
5
12165
XM_017005582.1
XP_016861071.1
4
986
XM_017005583.1
XP_016861072.1
4
436
XM_017005584.1
XP_016861073.1
4
986
XM_017005585.1
XP_016861074.1
4
986
XM_017005586.1
XP_016861075.1
4
442
XM_017005587.1
XP_016861076.1
4
605
XM_017005588.1
XP_016861077.1
5
1078
XM_017005589.1
XP_016861078.1
5
528
XM_017005590.1
XP_016861079.1
5
12165
XM_017005591.1
XP_016861080.1
4
986
XM_017005592.1
XP_016861081.1
4
986
XM_017005593.1
XP_016861082.1
4
532
XM_017005594.1
XP_016861083.1
4
436
XM_017005595.1
XP_016861084.1
5
1078
XM_017005596.1
XP_016861085.1
4
986
XM_017005597.1
XP_016861086.1
4
986
XM_017005598.1
XP_016861087.1
5
1078
XM_017005599.1
XP_016861088.1
4
986
XM_017005600.1
XP_016861089.1
4
605
XM_017005601.1
XP_016861090.1
4
436
XM_017005602.1
XP_016861091.1
4
986
XM_017005603.1
XP_016861092.1
4
605
XM_017005604.1
XP_016861093.1
4
436
XM_017005605.1
XP_016861094.1
5
1078
XM_017005606.1
XP_016861095.1
5
528
XM_017005607.1
XP_016861096.1
4
986
XM_017005608.1
XP_016861097.1
4
986
XM_017005609.1
XP_016861098.1
5
1078
XM_017005610.1
XP_016861099.1
4
986
XM_017005611.1
XP_016861100.1
4
986
XM_017005612.1
XP_016861101.1
4
986
GenBank mRNA
AF112220.1
-
3
386
AK055064.1
-
4
439
AK289868.1
-
4
433
AK294484.1
-
3
374
AV718064.1
-
4
430
AV718166.1
-
4
439
BC017805.1
-
3
421
BC036399.1
-
4
401
5263894
BC041385.1
-
4
1174
BC051828.1
-
4
413
6012704
BC066651.1
-
2
940
BG535216.1
-
3
422
BX488342.1
-
3
525
DA275579.1
-
3
527
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0