Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKAG2 SIRNA Assays ›
Gene Symbol:
PRKAG2
Gene Name:
protein kinase AMP-activated non-catalytic subunit gamma 2
Gene Aliases:
AAKG, AAKG2, CMH6, H91620p, WPWS
Chromosome Location:
Chr.7: 151556114 - 151877231 on Build GRCh38
UniGene ID:
Hs.647072
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24136
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKAG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040633.1
NP_001035723.1
7
1223
NM_001304527.1
NP_001291456.1
5
827
NM_001304531.1
NP_001291460.1
4
245
NM_016203.3
NP_057287.2
7
1492
NM_024429.1
NP_077747.1
3
317
XM_005250002.3
XP_005250059.1
7
1358
XM_005250004.3
XP_005250061.1
7
1304
XM_005250006.4
XP_005250063.1
5
1057
XM_006716021.2
XP_006716084.1
7
895
XM_011516282.1
XP_011514584.1
7
892
XM_011516283.1
XP_011514585.1
7
895
XM_011516284.1
XP_011514586.1
7
892
XM_011516285.1
XP_011514587.1
4
737
XM_011516286.1
XP_011514588.1
3
258
XM_011516287.1
XP_011514589.1
6
968
XM_017012268.1
XP_016867757.1
7
1301
XM_017012269.1
XP_016867758.1
7
1106
XM_017012270.1
XP_016867759.1
7
882
XM_017012271.1
XP_016867760.1
7
1301
XM_017012272.1
XP_016867761.1
7
879
XM_017012273.1
XP_016867762.1
5
1042
XM_017012274.1
XP_016867763.1
4
734
XM_017012275.1
XP_016867764.1
6
1231
XM_017012276.1
XP_016867765.1
7
1327
XM_017012277.1
XP_016867766.1
3
258
XM_017012278.1
XP_016867767.1
6
968
XM_017012279.1
XP_016867768.1
6
1234
XM_017012280.1
XP_016867769.1
6
1231
XM_017012281.1
XP_016867770.1
6
1257
XM_017012282.1
XP_016867771.1
5
677
GenBank mRNA
AB025580.1
-
3
230
AF087875.1
-
3
278
AJ249976.1
-
7
971
AK001887.1
-
3
317
AK301123.1
-
5
776
AK307798.1
-
6
1039
AK308116.1
-
7
923
AK314646.1
-
3
279
AW664063.1
-
4
217
BC020540.1
-
3
198
BC068598.1
-
7
1192
5259756
BT007127.1
-
3
158
DA151645.1
-
4
246
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_184051, XM_006235953, XM_008762653, XM_008762654, XM_006235951, and others.
Panther Classification:
Molecular Function -
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
glycogen metabolic process
regulation of glycolytic process
protein phosphorylation
negative regulation of protein kinase activity
fatty acid biosynthetic process
ATP biosynthetic process
carnitine shuttle
cell cycle arrest
positive regulation of peptidyl-threonine phosphorylation
sterol biosynthetic process
macroautophagy
regulation of fatty acid metabolic process
activation of protein kinase activity
intracellular signal transduction
regulation of fatty acid biosynthetic process
positive regulation of protein kinase activity
regulation of fatty acid oxidation
regulation of glucose import
negative regulation of protein serine/threonine kinase activity
regulation of signal transduction by p53 class mediator
AMP-activated protein kinase activity
cAMP-dependent protein kinase inhibitor activity
ATP binding
cAMP-dependent protein kinase regulator activity
phosphorylase kinase regulator activity
AMP binding
protein kinase binding
protein kinase activator activity
ADP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0