Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI2 SIRNA Assays ›
Gene Symbol:
ABI2
Gene Name:
abl-interactor 2
Gene Aliases:
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location:
Chr.2: 203327874 - 203432173 on Build GRCh38
UniGene ID:
Hs.471156
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241471
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282925.1
NP_001269854.1
10
1511
NM_001282926.1
NP_001269855.1
9
1493
NM_001282927.1
NP_001269856.1
8
986
NM_001282932.1
NP_001269861.1
9
1507
NM_005759.5
NP_005750.4
9
1397
XM_005246217.2
XP_005246274.1
11
1963
XM_005246218.2
XP_005246275.1
10
1945
XM_005246221.2
XP_005246278.1
9
1798
XM_005246225.2
XP_005246282.1
8
1711
XM_005246226.2
XP_005246283.1
8
1675
XM_006712174.2
XP_006712237.1
11
1963
XM_006712177.2
XP_006712240.1
12
2039
XM_006712179.2
XP_006712242.1
10
1816
XM_006712180.2
XP_006712243.1
10
1780
XM_006712181.2
XP_006712244.1
9
1693
XM_006712182.2
XP_006712245.1
9
1690
XM_006712185.1
XP_006712248.1
9
1751
XM_006712186.2
XP_006712249.1
7
1528
XM_011510472.2
XP_011508774.1
10
1813
XM_011510473.2
XP_011508775.1
10
1262
XM_017003118.1
XP_016858607.1
11
1960
XM_017003119.1
XP_016858608.1
11
1957
XM_017003120.1
XP_016858609.1
11
1960
XM_017003121.1
XP_016858610.1
10
1873
XM_017003122.1
XP_016858611.1
10
1810
XM_017003123.1
XP_016858612.1
9
1795
XM_017003124.1
XP_016858613.1
9
1762
XM_017003125.1
XP_016858614.1
9
1759
XM_017003126.1
XP_016858615.1
9
1756
XM_017003127.1
XP_016858616.1
9
1759
XM_017003128.1
XP_016858617.1
11
1952
XM_017003129.1
XP_016858618.1
9
1729
XM_017003130.1
XP_016858619.1
9
1726
XM_017003131.1
XP_016858620.1
8
1708
XM_017003132.1
XP_016858621.1
9
1693
XM_017003133.1
XP_016858622.1
11
1892
XM_017003134.1
XP_016858623.1
8
1675
XM_017003135.1
XP_016858624.1
8
1672
XM_017003136.1
XP_016858625.1
8
1672
XM_017003137.1
XP_016858626.1
11
1853
XM_017003138.1
XP_016858627.1
9
1630
XM_017003139.1
XP_016858628.1
10
1838
XM_017003140.1
XP_016858629.1
10
1835
XM_017003141.1
XP_016858630.1
8
1615
XM_017003142.1
XP_016858631.1
8
1612
XM_017003143.1
XP_016858632.1
10
1802
XM_017003144.1
XP_016858633.1
8
1546
XM_017003145.1
XP_016858634.1
7
1525
XM_017003146.1
XP_016858635.1
7
1528
GenBank mRNA
AF260261.1
-
10
1235
AK294429.1
-
4
563
AK297537.1
-
5
452
AK299092.1
-
8
729
AK299672.1
-
9
888
AK299679.1
-
4
429
AK299688.1
-
6
689
AK299750.1
-
4
335
AK299824.1
-
9
1337
AK302853.1
-
9
1433
AK316343.1
-
4
389
BC001439.2
-
9
1146
3138341
BT009920.1
-
9
1121
U23435.1
-
8
947
U31089.1
-
8
866
X95632.1
-
9
1221
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
cytoskeleton organization
actin polymerization or depolymerization
viral process
cell migration
Rac protein signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of Arp2/3 complex-mediated actin nucleation
DNA binding
protein binding
cytoskeletal adaptor activity
SH3 domain binding
kinase binding
ubiquitin protein ligase binding
protein complex binding
Rac GTPase binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0