Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241486
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122824.1
NP_001116296.1
3
650
NM_001146216.2
NP_001139688.1
5
1826
NM_006283.2
NP_006274.2
5
1880
XM_005273625.4
XP_005273682.1
7
2030
XM_005273628.1
XP_005273685.1
6
1537
XM_005273629.1
XP_005273686.1
7
1550
XM_005273630.1
XP_005273687.1
6
1558
XM_011544631.1
XP_011542933.1
6
1916
XM_011544632.2
XP_011542934.1
8
2091
XM_011544633.1
XP_011542935.1
6
1916
XM_011544634.1
XP_011542936.1
5
1880
XM_011544635.1
XP_011542937.1
6
1620
XM_011544636.2
XP_011542938.1
3
276
XM_011544637.1
XP_011542939.1
3
650
XM_011544638.2
XP_011542940.1
3
253
XM_017013766.1
XP_016869255.1
5
1501
XM_017013767.1
XP_016869256.1
6
1514
XM_017013768.1
XP_016869257.1
5
1501
XM_017013769.1
XP_016869258.1
6
1514
XM_017013770.1
XP_016869259.1
5
1450
XM_017013771.1
XP_016869260.1
5
1722
XM_017013772.1
XP_016869261.1
4
1414
XM_017013773.1
XP_016869262.1
4
1681
XM_017013774.1
XP_016869263.1
4
1366
XM_017013775.1
XP_016869264.1
5
1522
XM_017013776.1
XP_016869265.1
6
1514
XM_017013777.1
XP_016869266.1
4
1678
XM_017013778.1
XP_016869267.1
4
1377
GenBank mRNA
AB029026.1
-
4
450
AF049910.1
-
5
1821
AK294931.1
-
5
1481
AK295071.1
-
5
1428
AK295841.1
-
4
1417
AK303596.1
-
4
1312
AK304507.1
-
7
1713
AK304725.1
-
2
138
AK304774.1
-
3
200
AK308849.1
-
3
221
AK314620.1
-
5
1880
AY039239.1
-
4
190
AY072873.2
-
3
292
AY072874.1
-
3
281
AY072875.1
-
4
279
AY072876.2
-
5
1432
AY177411.1
-
3
591
BC041391.1
-
7
2077
5276767
BX647696.1
-
5
1825
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0