Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241488
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122824.1
NP_001116296.1
3
622
NM_001146216.2
NP_001139688.1
5
1798
NM_006283.2
NP_006274.2
5
1852
XM_005273625.4
XP_005273682.1
7
2002
XM_005273628.1
XP_005273685.1
6
1509
XM_005273629.1
XP_005273686.1
7
1522
XM_005273630.1
XP_005273687.1
6
1530
XM_011544631.1
XP_011542933.1
6
1888
XM_011544632.2
XP_011542934.1
8
2063
XM_011544633.1
XP_011542935.1
6
1888
XM_011544634.1
XP_011542936.1
5
1852
XM_011544635.1
XP_011542937.1
6
1592
XM_011544636.2
XP_011542938.1
3
248
XM_011544637.1
XP_011542939.1
3
622
XM_011544638.2
XP_011542940.1
3
225
XM_017013766.1
XP_016869255.1
5
1473
XM_017013767.1
XP_016869256.1
6
1486
XM_017013768.1
XP_016869257.1
5
1473
XM_017013769.1
XP_016869258.1
6
1486
XM_017013770.1
XP_016869259.1
5
1422
XM_017013771.1
XP_016869260.1
5
1694
XM_017013772.1
XP_016869261.1
4
1386
XM_017013773.1
XP_016869262.1
4
1653
XM_017013774.1
XP_016869263.1
4
1338
XM_017013775.1
XP_016869264.1
5
1494
XM_017013776.1
XP_016869265.1
6
1486
XM_017013777.1
XP_016869266.1
4
1650
XM_017013778.1
XP_016869267.1
4
1349
GenBank mRNA
AB029026.1
-
4
422
AF049910.1
-
5
1793
AK294931.1
-
5
1453
AK295071.1
-
5
1400
AK295841.1
-
4
1389
AK303596.1
-
4
1284
AK304507.1
-
7
1685
AK304725.1
-
2
110
AK304774.1
-
3
172
AK308849.1
-
3
193
AK314620.1
-
5
1852
AY039239.1
-
4
162
AY072873.2
-
3
264
AY072874.1
-
3
253
AY072875.1
-
4
251
AY072876.2
-
5
1404
AY177411.1
-
3
563
BC041391.1
-
7
2049
5276767
BX647696.1
-
5
1797
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0