Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other R3HCC1L SIRNA Assays ›
          Gene Symbol:
          R3HCC1L
          Gene Name:
          R3H domain and coiled-coil containing 1 like
          Gene Aliases:
          C10orf28, GIDRP86, GIDRP88, PSORT
          Chromosome Location:
          Chr.10: 98134608 - 98244897 on Build GRCh38
          UniGene ID:
          Hs.419800
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 241539

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: R3HCC1L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256619.1 NP_001243548.1 11 2738
          NM_001256620.1 NP_001243549.1 9 2645
          NM_001256621.1 NP_001243550.1 8 842
          NM_014472.4 NP_055287.4 9 2641
          NM_138469.2 NP_612478.2 10 2738
          XM_011539639.1 XP_011537941.1 11 2995
          XM_011539640.1 XP_011537942.1 12 2800
          XM_011539641.1 XP_011537943.1 11 2752
          XM_011539642.1 XP_011537944.1 12 3092
          XM_011539643.1 XP_011537945.1 10 2679
          XM_011539644.1 XP_011537946.1 10 2683
          XM_011539645.2 XP_011537947.1 10 2926
          XM_011539646.1 XP_011537948.1 9 2558
          XM_011539647.1 XP_011537949.1 9 2560
          XM_011539648.1 XP_011537950.1 8 2433
          XM_011539649.1 XP_011537951.1 10 2953
          XM_017016074.1 XP_016871563.1 10 3891
          XM_017016075.1 XP_016871564.1 7 2391
          XM_017016076.1 XP_016871565.1 10 2685
          XM_017016077.1 XP_016871566.1 12 2848
          XM_017016078.1 XP_016871567.1 11 2770
          XM_017016079.1 XP_016871568.1 11 2784
          XM_017016080.1 XP_016871569.1 9 2690
          XM_017016081.1 XP_016871570.1 10 2772
          XM_017016082.1 XP_016871571.1 8 2564
          XM_017016083.1 XP_016871572.1 9 2580
          GenBank mRNA AF525304.1 - 11 2691
          AK058187.1 - 5 432
          AK094479.1 - 6 1152
          AK292546.1 - 9 2640
          BC009231.2 - 5 567
          BC047908.1 - 8 788
          BC098175.1 - 6 585
          BC098268.1 - 10 2679 40026913
          BC098359.1 - 9 2582 40026912
          BC099848.1 - 9 2582 40026916

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-slzzl:80/100.66.73.10:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0