Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HELLS SIRNA Assays ›
          Gene Symbol:
          HELLS
          Gene Name:
          helicase, lymphoid-specific
          Gene Aliases:
          ICF4, LSH, Nbla10143, PASG, SMARCA6
          Chromosome Location:
          Chr.10: 94545767 - 94613905 on Build GRCh38
          UniGene ID:
          Hs.463677
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 241696

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HELLS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289067.1 NP_001275996.1 14, 15 1781
          NM_001289068.1 NP_001275997.1 13, 14 1598
          NM_001289069.1 NP_001275998.1 13, 14 1547
          NM_001289070.1 NP_001275999.1 11, 12 1349
          NM_001289071.1 NP_001276000.1 14, 15 1687
          NM_001289072.1 NP_001276001.1 11, 12 1253
          NM_001289073.1 NP_001276002.1 12, 13 1578
          NM_001289074.1 NP_001276003.1 14, 15 1562
          NM_001289075.1 NP_001276004.1 12, 13 1446
          NM_018063.4 NP_060533.2 13, 14 1643
          GenBank mRNA AB074174.1 - 1, 2 91
          AB102716.1 - 14, 15 1532
          AB102717.1 - 12, 13 1423
          AB102718.1 - 13, 14 1392
          AB102719.1 - 11, 12 1098
          AB102720.1 - 12, 13 1291
          AB102721.1 - 11, 12 1194
          AB102722.1 - 14, 15 1407
          AB102723.1 - 14, 15 1626
          AB113248.1 - 13, 14 1488
          AB113249.1 - 13, 14 1488
          AF155827.1 - 13, 14 1593
          AK314485.1 - 13, 14 1632
          AY007108.1 - 1, 2 100
          BC029381.1 - 1, 2 31
          BC030963.1 - 2, 3 246
          BC031004.1 - 2, 3 246
          BX538033.1 - 13, 14 1582

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          methylation-dependent chromatin silencing
          transcription, DNA-templated
          mitotic nuclear division
          multicellular organism development
          maintenance of DNA methylation
          pericentric heterochromatin assembly
          lymphocyte proliferation
          cell division
          helicase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0