Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R3 SIRNA Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
UniGene ID:
Hs.503022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241718
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164160.1
NP_001157632.1
17
2022
NM_001164161.1
NP_001157633.1
17
2022
NM_001164162.1
NP_001157634.1
17
2004
NM_001164163.1
NP_001157635.1
16
1935
NM_001164164.1
NP_001157636.1
16
1887
NM_018312.4
NP_060782.2
16
1887
XM_006718608.3
XP_006718671.1
18
2138
XM_006718612.3
XP_006718675.1
18
2120
XM_006718613.2
XP_006718676.1
18
2138
XM_006718614.2
XP_006718677.1
18
2120
XM_006718618.3
XP_006718681.1
17
2051
XM_006718621.2
XP_006718684.1
17
2000
XM_006718622.3
XP_006718685.1
16
1898
XM_006718623.3
XP_006718686.1
16
1881
XM_006718624.2
XP_006718687.1
16
1898
XM_011545135.1
XP_011543437.1
18
2138
XM_011545136.2
XP_011543438.1
17
2021
XM_011545137.2
XP_011543439.1
18
2121
XM_011545138.2
XP_011543440.1
19
2339
XM_011545139.1
XP_011543441.1
18
2120
XM_011545140.2
XP_011543442.1
18
2138
XM_011545141.1
XP_011543443.1
18
2087
XM_011545142.2
XP_011543444.1
17
2051
XM_011545143.1
XP_011543445.1
17
2033
XM_011545145.2
XP_011543447.1
17
1985
XM_017017962.1
XP_016873451.1
17
1976
XM_017017963.1
XP_016873452.1
18
2062
XM_017017964.1
XP_016873453.1
18
2120
XM_017017965.1
XP_016873454.1
17
2003
XM_017017966.1
XP_016873455.1
18
2206
XM_017017967.1
XP_016873456.1
18
2087
XM_017017968.1
XP_016873457.1
17
1970
XM_017017969.1
XP_016873458.1
18
2087
XM_017017970.1
XP_016873459.1
17
1970
XM_017017971.1
XP_016873460.1
17
2051
XM_017017972.1
XP_016873461.1
16
1907
XM_017017973.1
XP_016873462.1
17
2034
XM_017017974.1
XP_016873463.1
17
2033
XM_017017975.1
XP_016873464.1
17
2000
XM_017017976.1
XP_016873465.1
16
1817
XM_017017977.1
XP_016873466.1
17
1909
XM_017017978.1
XP_016873467.1
16
1760
XM_017017979.1
XP_016873468.1
16
1742
XM_017017980.1
XP_016873469.1
16
1760
XM_017017981.1
XP_016873470.1
16
1709
XM_017017982.1
XP_016873471.1
15
1673
XM_017017983.1
XP_016873472.1
17
2159
XM_017017984.1
XP_016873473.1
16
2002
GenBank mRNA
AB046778.2
-
17
1970
AF264779.1
-
15
1792
AF264780.1
-
15
1792
AK057250.1
-
11
1095
AK125055.1
-
2
1774
AK300359.1
-
9
856
AK302611.1
-
9
876
AK303315.1
-
12
1338
AK315136.1
-
16
1904
BC020953.1
-
3
144
BC105933.1
-
16
1890
40029064
BC105934.1
-
16
1890
40029058
BC107599.1
-
15
1803
40029065
CR749815.1
-
17
1967
CR933658.1
-
17
1985
CR936718.1
-
17
1990
DQ111954.2
-
15
1755
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0