Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: TSNAX-DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028393.1 | 6 | 1601 | |||
NR_028394.1 | 7 | 1729 | ||||
NR_028395.1 | 7 | 1729 | ||||
NR_028396.1 | 6 | 1601 | ||||
NR_028397.1 | 6 | 1466 | ||||
NR_028398.1 | 4 | 1220 | ||||
NR_028399.1 | 8 | 1836 | ||||
NR_028400.1 | 7 | 1708 |