Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC00477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029451.2 | 1 | 292 | |||
GenBank mRNA | AK057456.1 | - | 1 | 292 | ||
BC104436.1 | - | 1 | 272 | |||
BC104437.1 | - | 1 | 272 |