Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GUK1 SIRNA Assays ›
Gene Symbol:
GUK1
Gene Name:
guanylate kinase 1
Gene Aliases:
GMK
Chromosome Location:
Chr.1: 228139643 - 228148955 on Build GRCh38
UniGene ID:
Hs.376933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
242336
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GUK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000858.5
NP_000849.1
3
403
NM_001159390.1
NP_001152862.1
2
238
NM_001159391.1
NP_001152863.1
2
152
NM_001242839.1
NP_001229768.1
2
178
NM_001242840.1
NP_001229769.1
2
238
XM_005273103.3
XP_005273160.1
4
574
XM_005273104.4
XP_005273161.1
4
572
XM_005273105.3
XP_005273162.1
3
431
XM_005273106.2
XP_005273163.1
2
619
XM_011544174.2
XP_011542476.1
2
3002
XM_017001091.1
XP_016856580.1
4
575
XM_017001092.1
XP_016856581.1
3
910
XM_017001093.1
XP_016856582.1
3
370
XM_017001094.1
XP_016856583.1
2
208
XM_017001095.1
XP_016856584.1
3
367
XM_017001096.1
XP_016856585.1
2
207
GenBank mRNA
AK124677.1
-
666
AK125698.1
-
1
1819
AK303845.1
-
2
191
AL832986.1
-
1
741
BC006249.2
-
3
341
3946293
BC007369.2
-
1
55
BC009914.2
-
3
315
2821563
BC091480.1
-
2
158
30340102
BE780558.1
-
2
125
BI771824.1
-
2
178
BM790067.1
-
2
278
BU162698.1
-
219
BU194785.1
-
204
BU931096.1
-
195
CR456836.1
-
1
100
CR542006.1
-
1
100
L76200.1
-
1
100
U66895.1
-
1
106
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide metabolic process
dGDP biosynthetic process
nucleobase-containing small molecule interconversion
GDP-mannose metabolic process
glycoprotein transport
ATP metabolic process
GMP metabolic process
dGMP metabolic process
dATP metabolic process
GDP biosynthetic process
nucleotide phosphorylation
guanylate kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-52p4d:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0