Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACVR1 SIRNA Assays ›
          Gene Symbol:
          ACVR1
          Gene Name:
          activin A receptor type 1
          Gene Aliases:
          ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
          Chromosome Location:
          Chr.2: 157736446 - 157875880 on Build GRCh38
          UniGene ID:
          Hs.470316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 242344

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACVR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105.4 NP_001096.1 11 1925
          NM_001111067.2 NP_001104537.1 11 1744
          XM_005246939.3 XP_005246996.1 11 1796
          XM_005246940.3 XP_005246997.1 11 1764
          XM_006712825.3 XP_006712888.1 11 1877
          XM_011512106.2 XP_011510408.1 12 1844
          XM_011512107.2 XP_011510409.1 11 1816
          XM_011512108.2 XP_011510410.1 11 3223
          GenBank mRNA AK313630.1 - 11 1720
          BC033867.1 - 11 1925 5209711
          BC036748.1 - 11 1925
          KU177885.1 - 9 1495
          KU177886.1 - 8 1297
          KU177887.1 - 7 1021
          L02911.1 - 12 1835
          Z22534.1 - 10 1598

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          patterning of blood vessels
          in utero embryonic development
          gastrulation with mouth forming second
          mesoderm formation
          neural crest cell migration
          acute inflammatory response
          embryonic heart tube morphogenesis
          outflow tract septum morphogenesis
          atrioventricular valve morphogenesis
          mitral valve morphogenesis
          endocardial cushion morphogenesis
          endocardial cushion fusion
          atrial septum primum morphogenesis
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          germ cell development
          determination of left/right symmetry
          negative regulation of signal transduction
          positive regulation of pathway-restricted SMAD protein phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          regulation of ossification
          positive regulation of cell migration
          positive regulation of bone mineralization
          BMP signaling pathway
          activin receptor signaling pathway
          negative regulation of activin receptor signaling pathway
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          smooth muscle cell differentiation
          pharyngeal system development
          pathway-restricted SMAD protein phosphorylation
          ventricular septum morphogenesis
          cardiac muscle cell fate commitment
          BMP signaling pathway involved in heart development
          endocardial cushion cell fate commitment
          cellular response to glucocorticoid stimulus
          cellular response to BMP stimulus
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of determination of dorsal identity
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta receptor activity, type I
          protein binding
          ATP binding
          activin receptor activity, type I
          peptide hormone binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0