Search
Search

PASK| Gene Symbol: PASK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252119.1 | NP_001239048.1 | 18 | 4190 | ||
| NM_001252120.1 | NP_001239049.1 | 19 | 4322 | |||
| NM_001252122.1 | NP_001239051.1 | 18 | 4064 | |||
| NM_015148.3 | NP_055963.2 | 18 | 4169 | |||
| XM_005246991.1 | XP_005247048.1 | 15 | 3504 | |||
| XM_011510826.1 | XP_011509128.1 | 18 | 4145 | |||
| XM_011510828.1 | XP_011509130.1 | 19 | 4246 | |||
| XM_011510829.1 | XP_011509131.1 | 18 | 4187 | |||
| XM_011510830.1 | XP_011509132.1 | 18 | 4188 | |||
| XM_011510831.2 | XP_011509133.1 | 16 | 4352 | |||
| XM_011510832.2 | XP_011509134.1 | 16 | 4331 | |||
| XM_011510833.1 | XP_011509135.1 | 15 | 3525 | |||
| XM_011510834.1 | XP_011509136.1 | 11 | 3248 | |||
| XM_011510835.1 | XP_011509137.1 | 11 | 2838 | |||
| XM_017003634.1 | XP_016859123.1 | 18 | 4413 | |||
| XM_017003635.1 | XP_016859124.1 | 19 | 4225 | |||
| XM_017003636.1 | XP_016859125.1 | 18 | 4181 | |||
| GenBank mRNA | AF387103.1 | - | 18 | 4109 | ||
| AK026769.1 | - | 4 | 1818 | |||
| AK296945.1 | - | 15 | 3674 | |||
| AK302417.1 | - | 18 | 4020 | |||
| AK302561.1 | - | 15 | 3504 | |||
| BC020054.1 | - | 14 | 3344 | |||
| BC043495.1 | - | 12 | 2926 | |||
| BC063585.1 | - | 18 | 4112 | 4419998 | ||
| CR749231.1 | - | 19 | 4323 | |||
| D50925.2 | - | 18 | 4121 | |||
| U79240.1 | - | 9 | 1443 | |||