Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYO15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016239.3 | NP_057323.3 | 60 | 10093 | ||
XM_017024714.1 | XP_016880203.1 | 57 | 10393 | |||
XM_017024715.1 | XP_016880204.1 | 58 | 10456 | |||
GenBank mRNA | AF144094.1 | - | 60 | 10093 | ||
AK294036.1 | - | 18 | 1776 | |||
AK297179.1 | - | 11 | 1020 | |||
AK297641.1 | - | 6 | 685 | |||
AK298825.1 | - | 6 | 470 | |||
AK310232.1 | - | 4 | 318 | |||
AL713794.1 | - | 7 | 1205 | |||
BX538062.1 | - | 4 | 939 |