Search
Search

DCTN1| Gene Symbol: DCTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135040.2 | NP_001128512.1 | 22, 23 | 3290 | ||
| NM_001135041.2 | NP_001128513.1 | 20, 21 | 3013 | |||
| NM_001190836.1 | NP_001177765.1 | 22, 23 | 3233 | |||
| NM_001190837.1 | NP_001177766.1 | 24, 25 | 3329 | |||
| NM_004082.4 | NP_004073.2 | 25, 26 | 3350 | |||
| NM_023019.3 | NP_075408.1 | 20, 21 | 3013 | |||
| NR_033935.1 | 21, 22 | 3316 | ||||
| GenBank mRNA | AK297286.1 | - | 22, 23 | 2985 | ||
| AK314352.1 | - | 22, 23 | 3234 | |||
| AK315014.1 | - | 20, 21 | 2805 | |||
| BC071583.1 | - | 20, 21 | 2930 | 4817849 | ||
| BX647925.1 | - | 21, 22 | 3077 | |||
| BX647972.1 | - | 21, 22 | 3096 | |||
| X98801.1 | - | 23, 24 | 2987 | |||