Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COMT SIRNA Assays ›
          Gene Symbol:
          COMT
          Gene Name:
          catechol-O-methyltransferase
          Gene Aliases:
          HEL-S-98n
          Chromosome Location:
          Chr.22: 19941740 - 19969975 on Build GRCh38
          UniGene ID:
          Hs.370408
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 2453
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: COMT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000754.3 NP_000745.1 4, 5 740
          NM_001135161.1 NP_001128633.1 4, 5 698
          NM_001135162.1 NP_001128634.1 4, 5 715
          NM_007310.2 NP_009294.1 2, 3 471
          XM_011529886.1 XP_011528188.1 4, 5 645
          XM_017028594.1 XP_016884083.1 4, 5 964
          XM_017028595.1 XP_016884084.1 4, 5 908
          GenBank mRNA AK130031.1 - 5, 6 520
          AK290440.1 - 4, 5 678
          BC000419.2 - 4, 5 668 2964400
          BC005867.2 - 4, 5 668 2964400
          BC011935.2 - 3, 4 583 3503220
          BC100018.1 - 4, 5 730 6163659
          CR456422.1 - 3, 4 506
          CR456997.1 - 2, 3 491
          EF103918.1 - 2, 3 232
          EF103920.1 - 3, 4 221
          EF103921.1 - 3, 4 283
          EF103922.1 - 3, 4 520
          FJ224345.1 - 4, 5 668
          FJ410131.1 - 2, 3 341
          FJ410132.1 - 2, 3 491
          M58525.1 - 4, 5 695
          M65212.1 - 3, 4 569

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00241349_m1

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          female pregnancy
          learning
          short-term memory
          estrogen metabolic process
          response to organic cyclic compound
          cellular response to phosphate starvation
          methylation
          response to lipopolysaccharide
          developmental process
          negative regulation of renal sodium excretion
          neurotransmitter catabolic process
          dopamine catabolic process
          response to drug
          negative regulation of dopamine metabolic process
          response to pain
          multicellular organismal reproductive process
          negative regulation of smooth muscle cell proliferation
          positive regulation of homocysteine metabolic process
          regulation of sensory perception of pain
          magnesium ion binding
          protein binding
          methyltransferase activity
          O-methyltransferase activity
          catechol O-methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sxbfd:80/100.66.76.248:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0