Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SSX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005636.3 | NP_005627.1 | 3 | 144 | ||
NM_175729.1 | NP_783856.1 | 3 | 144 | |||
XM_017029759.1 | XP_016885248.1 | 3 | 202 | |||
GenBank mRNA | AX740218.1 | - | 2 | 86 | ||
BC005325.1 | - | 3 | 144 | 3961688 | ||
BC137394.1 | - | 2 | 111 | 9021392 | ||
BC137395.1 | - | 2 | 111 | 9021393 | ||
U90841.1 | - | 2 | 86 |
Gene Symbol: SSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278697.1 | NP_001265626.1 | 3 | 222 | ||
NM_003147.5 | NP_003138.3 | 3 | 222 | |||
NM_175698.2 | NP_783629.1 | 3 | 222 | |||
GenBank mRNA | BC002818.2 | - | 3 | 156 | 3636268 | |
BC007343.2 | - | 3 | 156 | 3615133 | ||
BC016957.1 | - | 3 | 156 | 3916128 | ||
BC069313.1 | - | 2 | 86 | 7262129 | ||
BC071827.1 | - | 3 | 156 | 4641362 | ||
BC103863.2 | - | 3 | 224 | 40003337 | ||
BG104299.1 | - | 3 | 194 | |||
BI560469.1 | - | 3 | 229 | |||
JX014236.1 | - | 2 | 86 | |||
X86175.1 | - | 3 | 177 | |||
Z49105.1 | - | 3 | 184 | |||
Z49106.1 | - | 2 | 116 |
Gene Symbol: SSX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164417.2 | NP_001157889.1 | 3 | 221 | ||
NM_001278701.1 | NP_001265630.1 | 3 | 221 | |||
NM_001278702.1 | NP_001265631.1 | 3 | 221 | |||
GenBank mRNA | BE391023.1 | - | 3 | 175 | ||
CB158955.1 | - | 3 | 138 |
Gene Symbol: SSX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021014.3 | NP_066294.1 | 3 | 226 | ||
XM_011543885.2 | XP_011542187.1 | 3 | 179 | |||
GenBank mRNA | AK310348.1 | - | 3 | 168 | ||
AK310513.1 | - | 3 | 179 | |||
BC005904.1 | - | 3 | 180 | 4064072 | ||
BC103862.1 | - | 3 | 225 | 40003336 | ||
DB444418.1 | - | 3 | 287 | |||
S82471.1 | - | 2 | 86 | |||
U90840.1 | - | 3 | 139 |
Gene Symbol: SSX4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034832.3 | NP_001030004.1 | 3 | 155 | ||
NM_001040612.2 | NP_001035702.1 | 3 | 155 | |||
XM_017029613.1 | XP_016885102.1 | 3 | 202 | |||
GenBank mRNA | AA312651.1 | - | 3 | 249 | ||
BE866727.1 | - | 3 | 125 |
Gene Symbol: SSX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021015.3 | NP_066295.3 | 4 | 262 | ||
NM_175723.1 | NP_783729.1 | 3 | 139 | |||
XM_011543949.2 | XP_011542251.1 | 3 | 174 | |||
XM_017029758.1 | XP_016885247.1 | 3 | 174 | |||
GenBank mRNA | BC016640.2 | - | 3 | 228 | 3923927 | |
BE896093.1 | - | 3 | 242 | |||
U90842.1 | - | 2 | 86 |
Gene Symbol: SSX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028366.1 | 3 | 158 | |||
GenBank mRNA | AK302765.1 | - | 3 | 150 |
Gene Symbol: SSX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173358.2 | NP_775494.1 | 3 | 245 | ||
XM_017029430.1 | XP_016884919.1 | 2 | 86 | |||
XM_017029431.1 | XP_016884920.1 | 2 | 86 |
Gene Symbol: SSX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073393.1 | 3 | 173 | |||
GenBank mRNA | DB029485.1 | - | 3 | 173 |