Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: VTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286371.1 | NP_001273300.1 | 7 | 2411 | ||
NM_001286372.1 | NP_001273301.1 | 6 | 2316 | |||
NM_016485.4 | NP_057569.2 | 8 | 2492 | |||
GenBank mRNA | AF271994.1 | - | 8 | 2381 | ||
AL136684.1 | - | 8 | 2385 | |||
BC022536.1 | - | 8 | 2391 | 4817057 |