Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ELOVL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104558.1 | NP_001098028.1 | 8 | 2887 | ||
NM_001297617.1 | NP_001284546.1 | 11 | 3285 | |||
NM_001297618.1 | NP_001284547.1 | 9 | 2892 | |||
NM_024930.2 | NP_079206.2 | 9 | 2938 | |||
XM_005248606.4 | XP_005248663.1 | 8 | 3502 | |||
XM_005248607.3 | XP_005248664.1 | 9 | 3215 | |||
XM_006714695.3 | XP_006714758.1 | 10 | 2994 | |||
XM_011543651.2 | XP_011541953.1 | 10 | 3184 | |||
XM_017009885.1 | XP_016865374.1 | 9 | 2875 | |||
XM_017009886.1 | XP_016865375.1 | 11 | 3114 | |||
XM_017009887.1 | XP_016865376.1 | 11 | 3305 | |||
GenBank mRNA | AB181393.1 | - | 9 | 2879 | ||
AL137506.1 | - | 4 | 2369 | |||
BC094792.1 | - | 11 | 3226 |
Gene Symbol: BICC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016677.1 | XP_016872166.1 | 1 | 4106 |
Gene Symbol: CNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093630.1 | - | 1 | 443 |
Gene Symbol: MINOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096240.1 | - | 1 | 523 |
Gene Symbol: NF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000268.3 | NP_000259.1 | 16 | 4484 | ||
NM_016418.5 | NP_057502.2 | 17 | 4529 | |||
NM_181828.2 | NP_861966.1 | 16 | 4403 | |||
NM_181829.2 | NP_861967.1 | 16 | 4406 | |||
NM_181830.2 | NP_861968.1 | 15 | 4280 | |||
NM_181832.2 | NP_861970.1 | 17 | 4544 | |||
NM_181833.2 | NP_861971.1 | 5 | 3194 | |||
XM_017028809.1 | XP_016884298.1 | 16 | 4047 | |||
XM_017028810.1 | XP_016884299.1 | 17 | 4088 | |||
GenBank mRNA | AF369657.1 | - | 16 | 4466 | ||
AF369658.1 | - | 17 | 4511 | |||
AK092306.1 | - | 1 | 1375 | |||
AK092515.1 | - | 1 | 226 |
Gene Symbol: PLEKHJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092742.1 | - | 2 | 1321 |
Gene Symbol: TYW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516372.2 | XP_011514674.1 | 16 | 3252 |
Gene Symbol: ZNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM993886.1 | - | 1 | 107 | ||
DB304820.1 | - | 1 | 89 |