Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 7 | 982 | ||
NM_182931.2 | NP_891847.1 | 8 | 1130 | |||
XM_005250493.1 | XP_005250550.1 | 7 | 1049 | |||
XM_011516400.1 | XP_011514702.1 | 9 | 1256 | |||
XM_017012435.1 | XP_016867924.1 | 8 | 1130 | |||
GenBank mRNA | AF067804.1 | - | 4 | 449 | ||
AF519459.1 | - | 7 | 797 | |||
AK000940.1 | - | 7 | 797 | |||
AK024088.1 | - | 2155 | ||||
AY147037.1 | - | 8 | 1130 | |||
AY157990.1 | - | 6 | 596 | |||
BC001296.2 | - | 8 | 1092 | |||
BC016382.1 | - | 4 | 295 | |||
BC024658.1 | - | 4 | 425 | |||
BC040004.1 | - | 7 | 1000 | |||
BC053906.1 | - | 7 | 1038 | |||
BC062583.1 | - | 8 | 1102 | |||
BC142987.1 | - | 7 | 1012 | |||
JQ809698.1 | - | 6 | 596 |